Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Acrolein

Acrolein, 5000 μg/ml in P and T methanol, 1 ml/ampoule, 1 Ampoul
  REST30645
 :   Restek
Acrolein
  REST30645
 :  Restek
 :  
Acrolein

Acrolein, 5000 μg/ml in P and T methanol, 1 ml/ampoule, 1 Ampoul

 :  
 
79,00 €
   
1 * 1 Ampull
 

 : This product has a limited shelf life. We recommend that you order only the ampoule quantity that meets your immediate needs.

 : NOTE: Store standards under frozen conditions (0 °C or colder). We recommend you store ampoules lying flat (horizontally) to prevent breakage. For best results, thaw the contents of the ampoule fully and shake well before opening and using the solution.

 

 

  • VE:
    1 Ampoul
  • Bezeichnung:
    Acrolein, 5000 μg/ml in P and T methanol
  • Pk Info:
    1 ml/ampoule