Isocyanates singles
4,4'-MDIP, 1000 µg/ml in dimethyl sulfoxide, 1 ml/ampoule, 1 Ampoul
Artikelnr. REST33003
Anbieter: Restek
Springe zu:
Product Details & Documents
OSHA 47 is for 4,4'-methylenediphenyl isocyanate (4,4'-MDI) in indoor air, and NIOSH Method 5522 is for 2,4-TDI; 2,6-TDI; 4,4'-MDI; and 1,6-hexamethylene diisocyanate (1,6-HDI) in air. Restek offers the 1,-(2-pyridyl)piperzine (1-2pp) derivative.
ASTM D5836 and OSHA 42 are test methods for determining 2,4-toluene diisocyanate (2,4-TDI) and 2,6-TDI in the workplace atmosphere.
Spezifikationen
Spezifikationen
- VE:1 Ampoul
- Bezeichnung:4,4'-MDIP, 1000 µg/ml in dimethyl sulfoxide
- Pk Info:1 ml/ampoule