Connectors, PP
Connector, straight, male luer lock/tubing nipple, 4,8 mm
Artikelnr. INFIPP-LMS48
Anbieter: PIEPER FILTER
Springe zu:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Spezifikationen
- Schlauch Innen-Ø:4,8 mm
- Bezeichnung:Connector, straight, male luer lock/tubing nipple
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.