Express Eingabe
Austria
AuftragsverfolgungKontakt
 

GC liners, LinerGOLD™

LinerGOLD splitless, single taper, 6,5 mm, 4,0 mm, 78,5
  HYPE453A2342-UI
GC liners, LinerGOLD™
  HYPE453A2342-UI
GC liners, LinerGOLD™
GC liners, LinerGOLD™

LinerGOLD splitless, single taper, 6,5 mm, 4,0 mm, 78,5

 
754,00 €
   
1 * 25 ST
 

LinerGOLD™ GC liners provide exceptional inertness, leading to increased accuracy and precision by preventing loss of sensitive compounds.

  • Lower detection limits for a wide range of active compounds
 
  • Ext.Ø:
    6,5 mm
  • Ø int.:
    4,0 mm
  • Bezeichnung:
    LinerGOLD splitless, single taper
  • Länge:
    78,5
  • Seq:
    30