Express Eingabe
Austria
AuftragsverfolgungKontakt
GC liners, LinerGOLD™

GC liners, LinerGOLD™

LinerGOLD straight, wool, 6,3, 4,0 mm, 78,5 mm
Catalog # HYPE453A1262-UI
GC liners, LinerGOLD™
Catalog # HYPE453A1262-UI
GC liners, LinerGOLD™
GC liners, LinerGOLD™

LinerGOLD straight, wool, 6,3, 4,0 mm, 78,5 mm

Price
608,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

LinerGOLD™ GC liners provide exceptional inertness, leading to increased accuracy and precision by preventing loss of sensitive compounds.

  • Lower detection limits for a wide range of active compounds
Specifications
  • Ext.Ø:
    6,3
  • Ø int.:
    4,0 mm
  • Bezeichnung:
    LinerGOLD straight, wool
  • Länge:
    78,5 mm
  • Seq:
    42
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.