MABSol® Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax® Human VEGF165, Tag Fr 1 * 25 µG
ACRBVE5-H8210-25UG
MABSol® Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax® Human VEGF165, Tag Fr 1 * 25 µG
ACRBVE5-H8210-25UG
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Aceton ≥99,5%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Schale, zum Kristallisieren
: VWR Collection
2-Propanol ≥99,8%, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Aceton ≥99,8%, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals