ContactUsLinkComponent
MULTIRAE LITE PUMPED O2/CO/H2S/NH3/LEL 1 * 1 ST
Artikelnr. RAESMAA301C128E010
Lieferant:  RAE SYSTEMS
MULTIRAE LITE PUMPED O2/CO/H2S/NH3/LEL 1 * 1 ST
Artikelnr. RAESMAA301C128E010
Lieferant:  RAE SYSTEMS
Frequently Bought Together