FLUORESCENT DYE 630-M STREPTAVIDIN FLUORESCENCE DYE 630-M WITH STREPTAVIDIN CONJUGATE. ALL THE FLUORESCENT DYE M SERIES PRODUCTS AR E DESIGNED TO BE MAXIMALLY EXCITED BY ONE OF THE MAJOR LIGHT SOURCES EQUIPPED IN FLOW CYTOMETERS. BESIDES, IN COMPARI 1 * 100 µG
Artikelnr. ABNOU0294
Lieferant: Abnova
Frequently Bought Together