ContactUsLinkComponent
FLUORESCENT DYE 488-I PHALLOIDIN FLUORESCENCE DYE 488-I WITH PHALLOIDIN CONJUGATE. FLUORESCENCE DYE 488-I CONJUGATES ARE SIGNIFICAN TLY BRIGHTER THAN FLUORESCEIN CONJUGATES AND MUCH MORE PHOTOSTABLE. ADDITIONALLY, THE FLUORESCENCE OF FLUORESCENCE DY 1 * 300 Assays
Artikelnr. ABNOU0281
Lieferant:  Abnova
FLUORESCENT DYE 488-I PHALLOIDIN FLUORESCENCE DYE 488-I WITH PHALLOIDIN CONJUGATE. FLUORESCENCE DYE 488-I CONJUGATES ARE SIGNIFICAN TLY BRIGHTER THAN FLUORESCEIN CONJUGATES AND MUCH MORE PHOTOSTABLE. ADDITIONALLY, THE FLUORESCENCE OF FLUORESCENCE DY 1 * 300 Assays
Artikelnr. ABNOU0281
Lieferant:  Abnova
Zugehörige Informationen
Frequently Bought Together