ContactUsLinkComponent
IL1RL1 (HUMAN) ELISA KIT IL1RL1 (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN IL1RL1 1 Kit 1 * 1 KIT
Artikelnr. ABNOKA4231
Lieferant:  Abnova
IL1RL1 (HUMAN) ELISA KIT IL1RL1 (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN IL1RL1 1 Kit 1 * 1 KIT
Artikelnr. ABNOKA4231
Lieferant:  Abnova
Frequently Bought Together