MB60 Wägebrücke lackiert Maximum load: 15kg / 30kg / 60kg Resolution: 5g, 10g, 20g Readability: 5g, 10g, 20g Platform Size (L x W): 500mm x 400mm Housing: die-cast aluminum Degree of protection: IP67 1 * 1 ST
Artikelnr. METB22006446
Lieferant: Mettler - Toledo
Frequently Bought Together