Dropping pipettes
Dropping pipettes with silicone teat, for 250 ml bottles, joint size 19/26
Artikelnr. BIBBSBD250
Anbieter: SciLabware
Springe zu:
Product Details & Documents
Soda-lime glass.
- With integral ground glass cone on pipette
- Supplied with rubber teats
- Bottles available separately
Spezifikationen
- Bezeichnung:Dropping pipettes with silicone teat, for 250 ml bottles, joint size 19/26
- Seq:03