AuftragsverfolgungKontakt
CF® Dye Dextran 10000 MW, anionic and fixable | Avantor
CF® Dye Dextran 10000 MW, anionic and fixable
CF568-Dextran 10000 MW, anionic and fixable, 562/583 nm, 1 mg
Artikelnr. 80113.
Anbieter:   Biotium
CF® Dye Dextran 10000 MW, anionic and fixable
Artikelnr. 80113.
Anbieter:  Biotium
CAS Nummer:  
CF® Dye Dextran 10000 MW, anionic and fixable
CF® Dye Dextran 10000 MW, anionic and fixable
CF568-Dextran 10000 MW, anionic and fixable, 562/583 nm, 1 mg
CAS Nummer:  
Preis
159,00 €
Einloggen für Verfügbarkeit
1 * 1 mg
Product Details & Documents
CF® dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular delivery. The dextran is 10000 MW, and contains a formaldehyde-fixable free-amine group.
  • Water-soluble fluid phase markers for tracking bulk endocytosis and cell permeability
  • Available with molecular weights of 3,5 kD, 10 kD, 40 kD, 70kD, 150 kD, or 250 kD
  • Choose from a selection of bright & photostable CF® dyes
Dextrans are water soluble branched-chain polysaccharides. Fluorescently labeled dextrans are used as markers for trafficking of fluid phase endocytic cargo to lysosomes. They are also used as tracers for epithelial and endothelial permeability. Labeled dextrans may also be used as microinjected tracers for neuronal morphology. CF® Dye Dextrans are available with a variety of dye colors and molecular weights ranging from 3500 to 250000.
These conjugates are available with Biotium's next-generation CF® dyes, a series of highly water-soluble fluorescent dyes spanning the visible and near-infrared spectrum for labeling biomolecules such as proteins and nucleic acids. Developed in house by Biotium's industry-leading chemists, CF® dyes offer superior brightness, photostability, and signal-to-noise when compared to other commercially available fluorescent dyes.
Spezifikationen
Spezifikationen
  • VE:
    1 mg
  • Bezeichnung:
    CF568-Dextran 10000 MW, anionic and fixable
  • Ex/Em:
    562/583 nm
Spezifikationen
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.