AuftragsverfolgungKontakt
VWR®, N-Plan Objektive
VWR®, N-Plan Objektive
N-Plan Objektiv, 60×/0,80 mm
Catalog # 630-2008
Supplier:   VWR Collection
VWR®, N-Plan Objektive
Catalog # 630-2008
Supplier:  VWR Collection
VWR®, N-Plan Objektive
VWR®, N-Plan Objektive
N-Plan Objektiv, 60×/0,80 mm
Price
159,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
Specifications
  • Vergrößerung:
    60×/0,80 mm
  • Bezeichnung:
    N-Plan Objektiv
  • Seq:
    211