Inlet liners
Liner, GC, ultra inert, straight, splitless, 2 mm
Artikelnr. HEWL5190-6168
Anbieter: Agilent
Springe zu:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Spezifikationen
- Ø int.:2 mm
- Bezeichnung:Liner, GC, ultra inert, straight, splitless
- Seq:03