Split/splitless inlet seals
Seal, gold plated, ultra inert, with washer
Artikelnr. HEWL5190-6145
Anbieter: Agilent
Springe zu:
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Lieferumfang: An inlet washer is provided with each inlet seal.
Spezifikationen
- Bezeichnung:Seal, gold plated, ultra inert, with washer
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.