Express Eingabe
Austria
AuftragsverfolgungKontakt
Galtek® check valves
Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" Quikgrip®, length 62,7 mm
Artikelnr. ENTECK4-4N-4Q
Anbieter:   ENTEGRIS
Galtek® check valves
Artikelnr. ENTECK4-4N-4Q
Anbieter:  ENTEGRIS
Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" Quikgrip®, length 62,7 mm
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
  • O-ring free design reduces contamination
  • All wetted parts are molded PFA and PTFE for corrosion resistance
  • No metal parts are used that could corrode or contribute to contamination
  • Poppet design allows for a high flow through rate and maximises rinsability
Spezifikationen
  • Bezeichnung:
    Inlet 1/4" FNPT, outlet 1/4" Quikgrip®, length 62,7 mm