Galtek® check valves
Inlet 1/2" FNPT, outlet 1/2" FNPT, length 84,3 mm
Artikelnr. ENTECK8-8N-8N
Anbieter: ENTEGRIS
Springe zu:
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
- O-ring free design reduces contamination
- All wetted parts are molded PFA and PTFE for corrosion resistance
- No metal parts are used that could corrode or contribute to contamination
- Poppet design allows for a high flow through rate and maximises rinsability
Spezifikationen
- Bezeichnung:Inlet 1/2" FNPT, outlet 1/2" FNPT, length 84,3 mm