AuftragsverfolgungKontakt
Boxes, with Hinged Lid
Boxes, with Hinged Lid
25×25×8 mm
Catalog # PLANG3950
Marketsource
Supplier:   PLANO W. PLANNET
Boxes, with Hinged Lid
Catalog # PLANG3950
Supplier:  PLANO W. PLANNET
Marketsource
Boxes, with Hinged Lid
Boxes, with Hinged Lid
25×25×8 mm
Price
113,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
These clear boxes are made of polystyrene.
  • With hinged lid
  • Good chemical resistance
  • Light and stackable
Multipurpose boxes for a wide range of storage and transport applications.
Specifications
  • Seq:
    01
  • B×T×H:
    25×25×8 mm