AuftragsverfolgungKontakt
Microscope objective lens mounts | Avantor
Microscope objective lens mounts
Lens holder with X/Y adjustment
Artikelnr. DIDAPOD060550
Anbieter:   DIDALAB
Microscope objective lens mounts
Artikelnr. DIDAPOD060550
Anbieter:  DIDALAB
Microscope objective lens mounts
Microscope objective lens mounts
Lens holder with X/Y adjustment
Preis
91,88 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The objective holder allows precise positioning of a microscope objective according to the vertical and the horizontal.
  • Both axes 2 mm each
  • Holder is mounted on a 10 mm diameter rod
Spezifikationen
  • Bezeichnung:
    Lens holder with X/Y adjustment
  • Seq:
    01