Koagulase-Test
Koagulase-Test, 10 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Nachweistest für Coagulasepositive Staphylococcus.
- Kalibriert gemäß ISO 6888-1
- Zum Differenzieren von Staphylococcus aureus und coagulasenegativen Staphylococci
Specifications
Specifications
- VE:10 mL
- Bezeichnung:Koagulase-Test
- Cat. No.:673030ZL
- Lagertemperatur:2...8 °C
- Shelf life:720
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.