Mobile sumps for storage containers
Mobile sump for drums and small containers, galvanized, for flammable and water hazardous liquids, with 60 l sump capacity and without grid, Verzinkter Stahl, 635×810×980 mm
MSGA23042
: asecos
Painted or galvanised steel.
- Easy to move by 2 swivel and 2 fixed wheels and push bar
For the storage of flammable and water hazardous liquids.
- Bezeichnung:Mobile sump for drums and small containers, galvanized, for flammable and water hazardous liquids, with 60 l sump capacity and without grid
- Material:Verzinkter Stahl
- Seq:02
- B×T×H:635×810×980 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.