Express Eingabe
Austria
AuftragsverfolgungKontakt
Mobile sumps for storage containers
Mobile sump for drums and small containers, painted, for flammable and water hazardous liquids, with 60 l sump capacity and without grid, 635×810×980 mm
Artikelnr. MSGA23041
Anbieter:   asecos
Mobile sumps for storage containers
Artikelnr. MSGA23041
Anbieter:  asecos
Mobile sumps for storage containers
Mobile sump for drums and small containers, painted, for flammable and water hazardous liquids, with 60 l sump capacity and without grid, 635×810×980 mm
Preis
412,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Painted or galvanised steel.
  • Easy to move by 2 swivel and 2 fixed wheels and push bar
For the storage of flammable and water hazardous liquids.
Spezifikationen
  • Bezeichnung:
    Mobile sump for drums and small containers, painted, for flammable and water hazardous liquids, with 60 l sump capacity and without grid
  • Seq:
    01
  • B×T×H:
    635×810×980 mm