Express Eingabe
Austria
AuftragsverfolgungKontakt
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Kit with HEPA filter and hose barb canister Ø 13 mm, for placing between the glove box and vacuum pump to protect the pump from particulate contaminants
Artikelnr. LBCP5441400
Anbieter:   LABCONCO
Accessories for glove boxes, Protector®
Artikelnr. LBCP5441400
Anbieter:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Kit with HEPA filter and hose barb canister Ø 13 mm, for placing between the glove box and vacuum pump to protect the pump from particulate contaminants
Preis
1 080,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Dokumente
Spezifikationen
  • Bezeichnung:
    Kit with HEPA filter and hose barb canister Ø 13 mm, for placing between the glove box and vacuum pump to protect the pump from particulate contaminants
  • Seq:
    20