Express Eingabe
Austria
AuftragsverfolgungKontakt
Accessories for glove boxes, Protector® | Avantor
Accessories for glove boxes, Protector®
Recirculation kit, incl. hose, clamps and epoxy-coated steel adapter, allows the built-in blower exhaust to be connected to the inlet HEPA/ ULPA filter
Artikelnr. LBCP5441100
Anbieter:   LABCONCO
Accessories for glove boxes, Protector®
Artikelnr. LBCP5441100
Anbieter:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Recirculation kit, incl. hose, clamps and epoxy-coated steel adapter, allows the built-in blower exhaust to be connected to the inlet HEPA/ ULPA filter
Preis
881,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Dokumente
Spezifikationen
  • Bezeichnung:
    Recirculation kit, incl. hose, clamps and epoxy-coated steel adapter, allows the built-in blower exhaust to be connected to the inlet HEPA/ ULPA filter
  • Seq:
    18