AMPSO ≥98%
Glasflasche für Feststoffe, 100 g
Purity: 98+% CAS No.: 68399-79-1 Molecular Formula: C7H17NO5S Formula Weight: 227.28 Physical Form: Crystals Appearance: White Melting Point: 215-225° Beilstein Reference: 5929916 MDL No.: MFCD00041777
Useful as a buffer
: N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
- :68399-79-1
- :C₇H₁₇NO₅S
- :MFCD00041777
- :227.28 g/mol
- :215…225 °C
- :Raumtemperatur
- :269-991-7
Documents
- Packung:Glasflasche für Feststoffe
- VE:100 g