0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
[EN]MIDIKROS 41,5CM 70KD MPES 1MM DRY 1 * 1 ST
SPECD04-E070-10-N
:
Spectrum Laboratories
[EN]MIDIKROS 41,5CM 70KD MPES 1MM DRY 1 * 1 ST
SPECD04-E070-10-N
:
Spectrum Laboratories
Frequently Bought Together
VWR®, Wägepapier, Sorte 531
: VWR Collection
Membranfilterscheiben, Supor® PES
: Cytiva (Formerly Pall Lab)
Quantitative Filterpapiere, aschefreie Sorten, Sorten 589/1 und 589/1 ½, Whatman™
: Whatman products (Cytiva)
Zubehör für MA-Serie
: Sartorius Balances
VWR®, Qualitative Filterpapiere
: VWR Collection
VWR®, Spritzenfilter
: VWR Collection
Filtertrichter, Sentino®
: Cytiva (Formerly Pall Lab)
VWR®, Vakuumfiltrationseinheiten, PES
: VWR Collection
VWR®, Glasfaserfilter
: VWR Collection
Glasmikrofaserfilter ohne Binder, Sorte GF/A, Whatman™
: Whatman products (Cytiva)
VWR®, Kryoboxen, Karton, 133×133 mm
: VWR Collection