ContactUsLinkComponent
with Software and Notebook Technical Specificaton: Worktable Capacity: 2 to 4 x 96 / 384 SBS PCR plates 2 to 4 x 96 tip rack (50/200 µl) Reagent Area 1: 8 x 1.5/2 ml microcentrifuge tube Reagent Area 2: 6 x 2 ml storage tube (free standing) and 1 x F 1 * 1 ST
Artikelnr. FTIT2200
Lieferant:  4titude Ltd.
with Software and Notebook Technical Specificaton: Worktable Capacity: 2 to 4 x 96 / 384 SBS PCR plates 2 to 4 x 96 tip rack (50/200 µl) Reagent Area 1: 8 x 1.5/2 ml microcentrifuge tube Reagent Area 2: 6 x 2 ml storage tube (free standing) and 1 x F 1 * 1 ST
Artikelnr. FTIT2200
Lieferant:  4titude Ltd.
Frequently Bought Together