Sample containers, straight, with screw cap, WHEATON®
150 ml, Printed label, sterile, Klar, PS/Metall
Artikelnr. 216-1016
Anbieter: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Artikelnr. 216-1016
Anbieter: DWK Life Sciences
Springe zu:
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Dokumente
Zertifikate
Suche nach ChargennummerCoC_LSUK360200_8321118 CoC_LSUK360206_0680228 CoC_LSUK360206_1580328 coc_LSUK360207_1050316 CoC_LSUK360207_3530708
Mehr
LSUK360251_TDS_EN
Technisches Datenblatt
Spezifikationen
- Typ:Printed label, sterile
- Kapazität:150 ml
- Farbe:Klar
- Material:PS/Metall