Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Vials, CrossLab

Vials, 0,25 ml, 12×32 mm, Nein, Screw
  HEWL8010-0194
 :   Agilent
Vials, CrossLab
  HEWL8010-0194
 :  Agilent
Vials, CrossLab

Vials, 0,25 ml, 12×32 mm, Nein, Screw

 
 
   
1 * 1 ST
 

These wide opening translucent vials are made from virgin PP and can be used with crimp or snap caps.

 : Meets the requirements of 21 CFR 177.1520

 
  • Mit Deckel:
    Nein
  • Ø×H:
    12×32 mm
  • Kapazität:
    0,25 ml
  • Bezeichnung:
    Vials
  • Seq:
    05
  • Verschlusstyp:
    Screw
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.