ContactUsLinkComponent
PROXIMA C16 PHI+ IMAGING SYSTEM Specifications: Resolution 1392 x 1040 pixels Exposure time 0.001 - 3600 s CCD cooling yes, triple Peltier and thermostated Camera bit depth 16 bit Electronic optical zoom 6 x Electronic iris control yes Electronic foc 1 * 1 ST
Artikelnr. IBSAIM-520-0750
Lieferant:  IBS Stock Items
PROXIMA C16 PHI+ IMAGING SYSTEM Specifications: Resolution 1392 x 1040 pixels Exposure time 0.001 - 3600 s CCD cooling yes, triple Peltier and thermostated Camera bit depth 16 bit Electronic optical zoom 6 x Electronic iris control yes Electronic foc 1 * 1 ST
Artikelnr. IBSAIM-520-0750
Lieferant:  IBS Stock Items
Frequently Bought Together