Express Eingabe
Austria
AuftragsverfolgungKontakt
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Glasflasche, 2,5 L
Catalog # 8088.2500
Flammable
Health
Irritant
Environment
Supplier:   Avantor
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Catalog # 8088.2500
Supplier:  Avantor
CAS Number:  110-54-3
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Glasflasche, 2,5 L

CAS Number:  110-54-3
Price
223,96 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specification Test Results

  • Assay (by GC):
    min. 99 %
  • Aromatic Compounds (as C6H6):
    max. 0,02 %
  • Boiling Range:
    max. 2°C
  • Density (g/ml) at 25°C:
    max. 0,657
  • Recorded Boiling Point:
    68.7°C
  • Residue after Evaporation:
    max. 5 ppm
  • Sulfur Compounds (as S):
    max. 0,005 %
  • Thiophene:
    passes test
  • Water (H2O):
    max. 0,02 %
  • Water Soluble Titrable Acid (meq/g):
    max. 0,0003
  • Trace Impurities (in ppm):
    -
  • Aluminium (Al):
    max. 0,5
  • Barium (Ba):
    max. 0,1
  • Boron (B):
    max. 0,02
  • Cadmium (Cd):
    max. 0,05
  • Calcium (Ca):
    max. 0,5
  • Chromium (Cr):
    max. 0,02
  • Cobalt (Co):
    max. 0,02
  • Copper (Cu):
    max. 0,02
  • Iron (Fe):
    max. 0,1
  • Lead (Pb):
    max. 0,1
  • Magnesium (Mg):
    max. 0,1
  • Manganese (Mn):
    max. 0,02
  • Nickel (Ni):
    max. 0,02
  • Tin (Sn):
    max. 0,1
  • Zinc (Zn):
    max. 0,1

Specifications

  • Packung:
    Glasflasche
  • VE:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.