Glyceryl Monooleate, Reference Standards
Glyceryl Monooleate 90%, USP reference standard, Vial, 250 mg
Artikelnr. 1295742.
Anbieter: USP
Springe zu:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Spezifikationen
Spezifikationen
- Packung:Vial
- VE:250 mg
- Normen:USP reference standard
- Bezeichnung:Glyceryl Monooleate 90%
- Cat. No.:1295742.
- CAS Nr.:25496-72-4
- Bezeichnung:Glyceryl Monooleate 90%
- Formel:C₂₁H₄₀O₄



