Heating tapes
Heating tape, 9,5 m length, 470 W
Artikelnr. WINKWBS00209500470
Anbieter: WINKLER
Springe zu:
Product Details & Documents
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
- Very high and constant heating power per meter
- High operating temperature
- Extremely flexible
- Quick and easy installation
Spezifikationen
- Bezeichnung:Heating tape, 9,5 m length, 470 W
- Seq:17