Heating tapes
Heating tape, 4,6 m length, 350 W
Artikelnr. WINKWBS00204600350
Anbieter: WINKLER
Springe zu:
Product Details & Documents
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
- Very high and constant heating power per meter
- High operating temperature
- Extremely flexible
- Quick and easy installation
Spezifikationen
- Bezeichnung:Heating tape, 4,6 m length, 350 W
- Seq:13
- Cat. No.:WINKWBS00204600350