AuftragsverfolgungKontakt
Pendulum hardness tester | Avantor
Pendulum hardness tester
König pendulum, stainless steel, with adjustable height, 200 g, SP0505
Artikelnr. SIMESP0505
Anbieter:   Industrial Physics
Pendulum hardness tester
Artikelnr. SIMESP0505
Anbieter:  Industrial Physics
Pendulum hardness tester
Pendulum hardness tester
König pendulum, stainless steel, with adjustable height, 200 g, SP0505
Preis
502,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Easy menu driven interface with jog dial
  • Design 'solid supports' to level the instrument
  • Integrated storage of glass plate and pendulum
  • Menu driven calibration menu eases both Persoz and König method
  • The end of a test is indicated by a visual and acoustic signal
The pendulum hardness test is based on the principle that the amplitude of oscillation of a pendulum touching a surface decreases more rapidly the softer the surface. It has a wide range of applications areas including laboratories, coating industry, paint industry, and the automotive industry.
Zertifizierung: ISO 1522 (2005) standard for the correct execution of the test.
Dokumente
Spezifikationen
  • Modell:
    SP0505
  • Bezeichnung:
    König pendulum, stainless steel, with adjustable height, 200 g
  • Seq:
    03