AuftragsverfolgungKontakt
Test charts | Avantor
Test charts
Test charts, Weiß/schwarz, 2DX
Artikelnr. SIMEVF3003
Anbieter:   Industrial Physics
Test charts
Artikelnr. SIMEVF3003
Anbieter:  Industrial Physics
Test charts
Test charts
Test charts, Weiß/schwarz, 2DX
Preis
184,00 €
Einloggen für Verfügbarkeit
1 * 500 ST
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Bestellinformation: A certificate is available for charts with an optical brightener.
Spezifikationen
  • Format:
    2DX
  • Farbe:
    Weiß/schwarz
  • Bezeichnung:
    Test charts
  • Seq:
    16