Test charts
Test charts, Weiß/schwarz, P121-10N
Artikelnr. SIMEVF3004
Anbieter: Industrial Physics
Springe zu:
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Bestellinformation: A certificate is available for charts with an optical brightener.
Dokumente
Spezifikationen
- Format:P121-10N
- Farbe:Weiß/schwarz
- Bezeichnung:Test charts
- Seq:20