Test charts
Test charts with optical brightner, Weiß/schwarz, A4, 210×297 mm
Artikelnr. SIMEVF2345
Anbieter: Industrial Physics
Springe zu:
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Bestellinformation: A certificate is available for charts with an optical brightener.
Dokumente
Spezifikationen
- Format:A4
- Farbe:Weiß/schwarz
- Bezeichnung:Test charts with optical brightner
- Seq:08
- B×H:210×297 mm