Express Eingabe
Austria
AuftragsverfolgungKontakt
Ethyllinoleat

Ethyllinoleat

Glass bottle for solids, 1 g
Catalog # CAYM10008198-1
Supplier:   Cayman Chemical
Ethyllinoleat
Catalog # CAYM10008198-1
Supplier:  Cayman Chemical
CAS Number:  544-35-4
Ethyllinoleat

Glass bottle for solids, 1 g

CAS Number:  544-35-4
Price
54,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  9Z,12Z-Octadecadienoic acid ethyl ester, Linoleic acid ethyl ester, cis,cis-9,12-Octadecadienoic acid ethyl ester, (9Z,12Z)-Ethyl octadeca-9,12-dienoate
  • CAS Number:544-35-4
  • Formula:C₂₀H₃₆O₂
  • MDL Number:MFCD00009535
  • Molecular Weight:308.5 g/mol
  • Storage Temperature:Gefrierschrank
  • EINECS:208-868-4
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.