Express Eingabe
Austria
AuftragsverfolgungKontakt
Racks for tubes, single use | Avantor

Racks for tubes, single use

13 mm, 100 (10×10), PS, 160×170×25 mm
Artikelnr. DELT901410
Anbieter:   DELTALAB
Racks for tubes, single use
Artikelnr. DELT901410
Anbieter:  DELTALAB
Racks for tubes, single use
Racks for tubes, single use

13 mm, 100 (10×10), PS, 160×170×25 mm

Preis
66,90 €
Einloggen für Verfügbarkeit
1 * 25 ST
Product Details & Documents

Polystyrene, flexible, white

Dokumente

Spezifikationen
  • Bohrung Ø:
    13 mm
  • Anz. Bohrungen:
    100 (10×10)
  • Material:
    PS
  • Seq:
    01
  • B×T×H:
    160×170×25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.