AuftragsverfolgungKontakt
TE Buffers 100X
TE Buffers 100X
TE buffer, 100X, 8,0, Plastic bottle, 500 mL
Catalog # 786-813
Supplier:   G-Biosciences
TE Buffers 100X
Catalog # 786-813
Supplier:  G-Biosciences
CAS Number:  
TE Buffers 100X
TE buffer, 100X, 8,0, Plastic bottle, 500 mL
CAS Number:  
Price
155,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.
Specifications
Specifications
  • Packung:
    Plastic bottle
  • VE:
    500 mL
  • Bezeichnung:
    TE buffer, 100X
  • pH-Wert:
    8,0
Specifications