COUNT-CAL PrecisionStandard 2µm, 6x25ml 1 * 150 mL
Catalog # MSPPCC02-PK
Supplier: MARKET SOURCE PART PROCESS
COUNT-CAL PrecisionStandard 2µm, 6x25ml 1 * 150 mL
Catalog # MSPPCC02-PK
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® AVS TITRINORM Pufferlösungen, 20 °C
Supplier: VWR Chemicals
Pufferlösungen, Certipur®, farbcodiert, Supelco®
Supplier: Merck
Pufferlösungen in Beuteln, Certipur® , Supelco®
Supplier: Merck
pH Pufferlösungen, DuraCal
Supplier: HAMILTON BONADUZ
Leitfähigkeitsstandardlösungen
Supplier: VWR Chemicals
Kaliumchlorid 3 mol/l (3 N) in wässriger Lösung, Supelco®
Supplier: Merck
Pufferlösung, Ready to Use
Supplier: WTW
DISTRILAB Count-Cal Count Precision Standard (6x25ml) 1 * 6 ST
Supplier: MARKET SOURCE PART PROCESS