Accessories for glove boxes, Precise™
Oxygen monitor, dor detecting trace oxygen levels using an electrochemical sensor; digital readout in three measurement ranges of 0-100 ppm, 100-1000 ppm and 1000-10000 ppm, for 230 V units
Artikelnr. LBCP5244501
Anbieter: LABCONCO
Springe zu:
Product Details & Documents
Spezifikationen
- Bezeichnung:Oxygen monitor, dor detecting trace oxygen levels using an electrochemical sensor; digital readout in three measurement ranges of 0-100 ppm, 100-1000 ppm and 1000-10000 ppm, for 230 V units
- Seq:16