AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Oxygen monitor for 115/230 V, measures percent of oxygen (0,1 - 100 %) inside the glove box using a chemically resistant electrochemical sensor, incl. interconnecting sensor cable and power cord for connection to the glove box’s interior electrical receptacle
Catalog # LBCP5244400
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5244400
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen monitor for 115/230 V, measures percent of oxygen (0,1 - 100 %) inside the glove box using a chemically resistant electrochemical sensor, incl. interconnecting sensor cable and power cord for connection to the glove box’s interior electrical receptacle
Price
4 040,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Bezeichnung:
    Oxygen monitor for 115/230 V, measures percent of oxygen (0,1 - 100 %) inside the glove box using a chemically resistant electrochemical sensor, incl. interconnecting sensor cable and power cord for connection to the glove box’s interior electrical receptacle
  • Seq:
    17