Vakuwash For rinsing and drying glass cuvettes. Cell is held, open end down, against a rubber cushion which closes vacuum system. Includes 500 mL PP flask, Vikem® stopper, tubing, and Vakuwash funnel. 1 * 1 ST
Artikelnr. SIALZ177474-1EA
Lieferant: Merck
Frequently Bought Together