Microplates, 96-well
96-Well-Platte, V-Boden, steril, Klar, 288 µl, 96
KART2622/S
: KARTELL
Multiple-well plates (96 sample wells) in clear PS. Available in three shapes: flat bottom, U bottom and V bottom wells.
- Aseptic or sterile
- Single-wrapped
Ordering information: PS covers are sold separately.
- Anz. Wells:96
- Volumen:288 µl
- Farbe:Klar
- Bezeichnung:96-Well-Platte, V-Boden, steril