Operating scissors
Straight, sharp/sharp, 155 mm
Artikelnr. HAMML-HSB104-15
Anbieter: HAMMACHER KARL
Springe zu:
Product Details & Documents
These operating scissors made from stainless steel are suitable for medical and clinical applications.
Spezifikationen
- Typ:Straight, sharp/sharp
- Länge:155 mm
- Seq:10