AuftragsverfolgungKontakt
Flasks, flat bottom with Quickfit® ground joints | Avantor
Flasks, flat bottom with Quickfit® ground joints
1000 ml, 190 mm, 24/29 NS
Artikelnr. BIBBFF1L/3M
Anbieter:   SciLabware
Flasks, flat bottom with Quickfit® ground joints
Artikelnr. BIBBFF1L/3M
Anbieter:  SciLabware
Flasks, flat bottom with Quickfit® ground joints
Flasks, flat bottom with Quickfit® ground joints
1000 ml, 190 mm, 24/29 NS
Preis
324,00 €
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
PYREX®, borosilicate glass, clear, with medium necks.
  • Very good chemical resistance
  • High temperature resistance
  • With longer neck than standard of flasks
Flat base allows use with hotplates. With labelling area.
Spezifikationen
  • Hülse:
    24/29 NS
  • Kapazität:
    1000 ml
  • Höhe:
    190 mm