Express Eingabe
Austria
AuftragsverfolgungKontakt
Gastro-norm container
Gastro-norm container
28 l, 530x325x200 mm
Catalog # HUPF0130652H
Supplier:   HUPFER METALLWERKE
Gastro-norm container
Catalog # HUPF0130652H
Supplier:  HUPFER METALLWERKE
Gastro-norm container
Gastro-norm container
28 l, 530x325x200 mm
Price
209,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10.
  • Fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose containers for a wide range of uses.
Specifications
  • Kapazität:
    28 l
  • Seq:
    02
  • Außen B×T×H:
    530x325x200 mm